Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxy-4'-methoxyflavone

Supplier: APOLLO SCIENTIFIC

OR14402-1G OR14402-25G OR14402-5G
77404-940EA 41.78 USD InStock
77404-940 77404-942 77404-946
3-Hydroxy-4'-methoxyflavone
3-Hydroxy-4'-methoxyflavone


Formula: C₁₆H₁₂O₄
MW: 268.27
Melting Pt: 234 °C
Storage Temperature: Ambient
MDL Number: MFCD00017682
CAS Number: 6889-78-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty