Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR1258-100G OR1258-25G
77404-374EA 234.32 USD InStock
77404-374 77404-376
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane 98%
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane


Formula: C₁₂H₂₈Cl₂OSi₂
MW: 315.43 g/mol
Boiling Pt: 70 °C/0.5 mmHg
Density: 0.986 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009655
CAS Number: 69304-37-6
UN: 2987
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty