Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-o-cresol 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR1558-25G OR1558-5G OR1558-100G
77405-474EA 221.08 USD InStock
77405-474 77405-476 77405-472
3-Chloro-o-cresol 98%
3-Chloro-o-cresol


Formula: C₇H₇ClO
MW: 142.58
Boiling Pt: 225 °C
Melting Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00079738
CAS Number: 3260-87-5
EINECS: 221-861-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty