Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dichloroisonicotinic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR1745-1G OR1745-25G OR1745-5G
77406-006EA 21.26 USD InStock
77406-006 77406-008 77406-010
3,5-Dichloroisonicotinic acid 98%
3,5-Dichloroisonicotinic acid


Formula: C₆H₃Cl₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD01861975
CAS Number: 13958-93-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty