Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-5-nitroindole-2-carboxylate 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR2221-10G OR2221-5G
77406-834EA 69.28 USD InStock
77406-834 77406-842
Ethyl-5-nitroindole-2-carboxylate 98%
Ethyl-5-nitroindole-2-carboxylate


Formula: C₁₁H₁₀N₂O₄
Melting Pt: 223…225 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00216477
CAS Number: 16732-57-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty