Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 4-((methylsulfonyl)oxy)piperidine-1-carboxylate 98%

Supplier: APOLLO SCIENTIFIC

OR16622-10G OR16622-25G
77405-786EA 50.94 USD InStock
77405-786 77405-788
tert-Butyl 4-((methylsulfonyl)oxy)piperidine-1-carboxylate 98%
tert-Butyl 4-((methylsulfonyl)oxy)piperidine-1-carboxylate


Formula: C₁₁H₂₁NO₅S
MW: 279.3500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04116220
CAS Number: 141699-59-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty