Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Chloroethyl chloroformate 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR16433-100G OR16433-25G OR16433-500G
77405-686EA 63.16 USD InStock
77405-686 77405-688 77405-690
1-Chloroethyl chloroformate 98%
1-Chloroethyl chloroformate


Formula: C₃H₄Cl₂O₂
MW: 142.97 g/mol
Boiling Pt: 118…121 °C
Melting Pt: -65
Density: 1.325 g/cm³ (25 °C)
Flash Pt: 40.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000647
CAS Number: 50893-53-3
EINECS: 256-834-2
UN: 2742
ADR: 6.1+3+8 II
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty