Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-methylimidazo[1,2-a]pyridine

Supplier: APOLLO SCIENTIFIC
Warning

OR18106-100MG OR18106-1G OR18106-250MG
77406-186EA 75.4 USD InStock
77406-186 77406-188 77406-190
5-Bromo-2-methylimidazo[1,2-a]pyridine
5-Bromo-2-methylimidazo[1,2-a]pyridine


Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10699211
CAS Number: 74420-51-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty