Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Chloronaphthalene-1-sulfonic acid 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR21393-1G OR21393-250MG OR21393-5G
77406-682EA 55.02 USD InStock
77406-682 77406-684 77406-686
8-Chloronaphthalene-1-sulfonic acid 97%
8-Chloronaphthalene-1-sulfonic acid


Formula: C₁₀H₇ClO₃S
MW: 242.679 g/mol
Melting Pt: 145…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00052379
CAS Number: 145-74-4
EINECS: 205-661-0
UN: 3261
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty