Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-4-(methylamino)piperidine

Supplier: APOLLO SCIENTIFIC
Danger

OR15676-100G OR15676-25G
77405-534EA 234.32 USD InStock
77405-534 77405-536
1-Boc-4-(methylamino)piperidine
1-Boc-4-(methylamino)piperidine


Formula: C₁₁H₂₂N₂O₂
MW: 214.3046 g/mol
Density: 1.005 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02259411
CAS Number: 147539-41-1
UN: 2811
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty