Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Nitro-4-picoline

Supplier: APOLLO SCIENTIFIC
Warning

OR16077-1G OR16077-5G
77405-622EA 588.13 USD InStock
77405-622 77405-624 77405-626
2-Nitro-4-picoline
2-Nitro-4-picoline


Formula: C₆H₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD04114155
CAS Number: 18368-71-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty