Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrahydrofuran-3-carbonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

OR16566-100MG OR16566-1G OR16566-250MG
77405-736EA 69.28 USD InStock
77405-736 77405-738 77405-740
Tetrahydrofuran-3-carbonyl chloride
Tetrahydrofuran-3-carbonyl chloride


Formula: C₅H₇ClO₂
Boiling Pt: 75…76 °C/15 mmHg
Storage Temperature: Ambient
MDL Number: MFCD06656696
CAS Number: 69595-02-4
UN: 3265
ADR: 8,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty