Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Iodobenzenesulfonic acid 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR16595-1G OR16595-25G OR16595-5G
77405-748EA 28.52 USD InStock
77405-748 77405-750 77405-752
2-Iodobenzenesulfonic acid 98%
2-Iodobenzenesulfonic acid


Formula: C₆H₅IO₃S
Storage Temperature: Ambient
MDL Number: MFCD01860232
CAS Number: 63059-25-6
UN: 3261
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty