Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-1H-pyrazolo[4,3-b]pyridine 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR17930-10G OR17930-1G OR17930-5G
77406-162EA 477.82 USD InStock
77406-162 77406-164 77406-166
3-Bromo-1H-pyrazolo[4,3-b]pyridine 98%
3-Bromo-1H-pyrazolo[4,3-b]pyridine


Formula: C₆H₄BrN₃
Storage Temperature: Ambient
MDL Number: MFCD11518974
CAS Number: 633328-33-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty