Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethylbenzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR21929-100G OR21929-250G OR21929-25G
77406-738EA 27.5 USD InStock
77406-738 77406-740 77406-742
4-Ethylbenzoic acid
4-Ethylbenzoic acid


Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 270
Melting Pt: 112…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002570
CAS Number: 619-64-7
EINECS: 210-605-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty