Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-1H-indole, N-BOC protected

Supplier: APOLLO SCIENTIFIC
Warning

OR17607-1G OR17607-250MG OR17607-5G
77406-052EA 186.52 USD InStock
77406-052 77406-054 77406-056
3-Methyl-1H-indole, N-BOC protected
3-Methyl-1H-indole, N-BOC protected


Formula: C₁₄H₁₇NO₂
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD13563142
CAS Number: 89378-43-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty