Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloro-5-methylpyrimidine 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR17853-100G OR17853-25G OR17853-5G
77406-122EA 58.08 USD InStock
77406-122 77406-124 77406-126
2,4-Dichloro-5-methylpyrimidine 98%
2,4-Dichloro-5-methylpyrimidine


Formula: C₅H₄Cl₂N₂
MW: 163.01
Boiling Pt: 235
Melting Pt: 26
Density: 1.39 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00023197
CAS Number: 1780-31-0
EINECS: 217-227-8
UN: 3261
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty