Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sulfamoyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

OR18738-1G OR18738-5G
77406-378EA 21.32 USD InStock
77406-378 77406-380
Sulfamoyl chloride
Sulfamoyl chloride


Formula: ClH₂NO₂S
MW: 115.54 g/mol
Melting Pt: 40 °C
Storage Temperature: Freezer
MDL Number: MFCD11519990
CAS Number: 7778-42-9
UN: 3261
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty