Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-thenoic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR22424-100G OR22424-25G OR22424-5G
77406-888EA 155.88 USD InStock
77406-888 77406-890 77406-892
5-Bromo-2-thenoic acid 97%
5-Bromo-2-thenoic acid


Formula: C₅H₃BrO₂S
Melting Pt: 138…144 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00079725
CAS Number: 7311-63-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty