Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bromocyclopropane 99+%

Supplier: APOLLO SCIENTIFIC
Danger

OR18407-100G OR18407-25G
77406-322EA 69.28 USD InStock
77406-322 77406-324
Bromocyclopropane 99+%
Bromocyclopropane


Formula: C₃H₅Br
Boiling Pt: 69 °C
Density: 1.51 g/cm³ (25 °C)
Flash Pt: –6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001271
CAS Number: 4333-56-6
EINECS: 224-375-7
UN: 1993
ADR: 3,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty