Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1,5-Dimethyl-1H-pyrazol-3-yl)methanol 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR23190-1G OR23190-5G
77407-164EA 68.26 USD InStock
77407-164 77407-166
(1,5-Dimethyl-1H-pyrazol-3-yl)methanol 97%
(1,5-Dimethyl-1H-pyrazol-3-yl)methanol


Formula: C₆H₁₀N₂O
Boiling Pt: 118 °C/2 mmHg
Melting Pt: 50…51 °C
Storage Temperature: Ambient
MDL Number: MFCD03659702
CAS Number: 153912-60-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty