Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Chloromethyl)benzoic acid

Supplier: APOLLO SCIENTIFIC
Danger

OR2365-25G OR2365-5G
77407-358EA 23.32 USD InStock
77407-358 77407-360
4-(Chloromethyl)benzoic acid
4-(Chloromethyl)benzoic acid


Formula: C₈H₇ClO₂
MW: 170.60 g/mol
Melting Pt: 201…202 °C
Storage Temperature: Ambient
MDL Number: MFCD00002568
CAS Number: 1642-81-5
EINECS: 216-697-1
UN: 3261
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty