Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Piperazin-1-ylnicotinonitrile 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR2418-1G OR2418-5G
77407-442EA 93.74 USD InStock
77407-442 77407-444
6-Piperazin-1-ylnicotinonitrile 95%
6-Piperazin-1-ylnicotinonitrile


Formula: C₁₀H₁₂N₄
MW: 188.23 g/mol
Melting Pt: 75…78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00833024
CAS Number: 149554-29-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty