Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromoaniline 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR2121-100G OR2121-25G OR2121-500G
77406-642EA 61.12 USD InStock
77406-642 77406-644 77406-646
3-Bromoaniline 98%
3-Bromoaniline


Formula: C₆H₆BrN
Boiling Pt: 74…75 °C/1 mmHg
Melting Pt: 15…18 °C
Density: 1.58 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007757
CAS Number: 591-19-5
EINECS: 209-704-4
UN: 2810
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty