Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-Ethyl-2-bromopropionate

Supplier: APOLLO SCIENTIFIC
Danger

OR2205-10KG OR2205-500G
77406-788EA 496.16 USD InStock
77406-788 77406-790
(±)-Ethyl-2-bromopropionate
(±)-Ethyl-2-bromopropionate


Formula: C₅H₉BrO₂
MW: 181.03 g/mol
Boiling Pt: 156…160 °C
Density: 1.394 g/cm³ (25 °C)
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00000144
CAS Number: 535-11-5
EINECS: 208-609-2
UN: 2920
ADR: 8+3 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty