Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dimethylthiazole

Supplier: APOLLO SCIENTIFIC
Warning

OR322034-100G OR322034-500G
77409-138EA 36.4 USD InStock
77409-138 77409-140
2,4-Dimethylthiazole
2,4-Dimethylthiazole


Formula: C₅H₇NS
Boiling Pt: 144…146 °C
Density: 1.05 g/cm³ (25 °C)
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00014509
CAS Number: 541-58-2
UN: 1993
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty