Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-1-(phenylsulfonyl)indole

Supplier: APOLLO SCIENTIFIC
Warning

OR23046-1G OR23046-25G OR23046-5G
77407-038EA 21.34 USD InStock
77407-038 77407-040 77407-042
3-Bromo-1-(phenylsulfonyl)indole
3-Bromo-1-(phenylsulfonyl)indole


Formula: C₁₄H₁₀BrNO₂S
MW: 336.203 g/mol
Melting Pt: 122…126 °C
Storage Temperature: Ambient
MDL Number: MFCD02681986
CAS Number: 99655-68-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty