Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-5-methylisoxazole-3-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

OR23145-100G OR23145-25G OR23145-5G
77407-136EA 227.2 USD InStock
77407-136 77407-138 77407-140
Methyl-5-methylisoxazole-3-carboxylate
Methyl 5-methylisoxazole-3-carboxylate


Formula: C₆H₇NO₃
MW: 141.13 g/mol
Boiling Pt: 125 °C/11 mmHg
Melting Pt: 91…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00015895
CAS Number: 19788-35-3
EINECS: 243-311-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty