Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Chloromethyl)benzoic acid 99%

Supplier: APOLLO SCIENTIFIC
Danger

OR2330-1G OR2330-25G OR2330-5G
77407-228EA 23.44 USD InStock
77407-228 77407-230 77407-232
2-(Chloromethyl)benzoic acid 99%
2-(Chloromethyl)benzoic acid


Formula: C₈H₇ClO₂
Melting Pt: 135 °C
Storage Temperature: Freezer
MDL Number: MFCD06208434
CAS Number: 85888-81-9
UN: 3261
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty