Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[2-(4-Morpholinyl)phenyl]methanamine 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR23385-1G OR23385-5G
77407-302EA 130.4 USD InStock
77407-302 77407-304
[2-(4-Morpholinyl)phenyl]methanamine 97%
[2-(4-Morpholinyl)phenyl]methanamine


Formula: C₁₁H₁₆N₂O
Storage Temperature: Ambient
MDL Number: MFCD03086183
CAS Number: 204078-48-8
UN: 2735
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty