Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(1,3-Oxazol-5-yl)thiophene-2-sulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

OR23225-1G OR23225-250MG
77407-190EA 261.84 USD InStock
77407-190 77407-192
5-(1,3-Oxazol-5-yl)thiophene-2-sulfonyl chloride
5-(1,3-Oxazol-5-yl)thiophene-2-sulfonyl chloride


Formula: C₇H₄ClNO₃S₂
Melting Pt: 80…82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02681961
CAS Number: 321309-40-4
UN: 1759
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty