Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Mucic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR2356-100G OR2356-1KG OR2356-500G
77407-338EA 21.36 USD InStock
77407-338 77407-340 77407-342
Mucic acid 97%
Mucic acid


Formula: C₆H₁₀O₈
Melting Pt: 210…230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004239
CAS Number: 526-99-8
EINECS: 208-404-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty