Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Pyrazinedicarboxylic anhydride 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR2581-25G OR2581-5G
77407-664EA 23.42 USD InStock
77407-664 77407-666
2,3-Pyrazinedicarboxylic anhydride 98%
2,3-Pyrazinedicarboxylic anhydride


Formula: C₆H₂N₂O₃
MW: 150.09 g/mol
Melting Pt: 210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00179418
CAS Number: 4744-50-7
EINECS: 225-260-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty