Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-1-bromocyclobutanecarboxylate 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR300697-1G OR300697-25G OR300697-5G
77408-692EA 21.32 USD InStock
77408-692 77408-694 77408-696
Ethyl-1-bromocyclobutanecarboxylate 97%
Ethyl-1-bromocyclobutanecarboxylate


Formula: C₇H₁₁BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00001322
CAS Number: 35120-18-4
UN: 3265
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty