Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,8-Dichloro-2-methylquinoline 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR307936-1G OR307936-250MG
77408-948EA 121.24 USD InStock
77408-948 77408-950
4,8-Dichloro-2-methylquinoline 97%
4,8-Dichloro-2-methylquinoline


Formula: C₁₀H₇Cl₂N
Storage Temperature: Ambient
CAS Number: 75896-69-4
UN: 2811
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty