Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-3-nitrobenzene 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR4281-100G OR4281-1G OR4281-500G
77409-680EA 96.78 USD InStock
77409-680 77409-682 77409-684
1-Bromo-3-nitrobenzene 98%
1-Bromo-3-nitrobenzene


Formula: C₆H₄BrNO₂
MW: 202.01
Boiling Pt: 256
Melting Pt: 51…54 °C
Density: 1.704 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00024298
CAS Number: 585-79-5
EINECS: 209-563-9
UN: 3459
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty