Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butylhydrazine hydrochloride

Supplier: APOLLO SCIENTIFIC
Warning

OR28606-1KG OR28606-500G
77408-114EA 115.12 USD InStock
77408-114 77408-116
tert-Butylhydrazine hydrochloride
tert-Butylhydrazine hydrochloride


Formula: C₄H₁₃ClN₂
MW: 124.61 g/mol
Melting Pt: 191…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00012947
CAS Number: 7400-27-3
EINECS: 231-001-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty