Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dibromo-1H-pyrazolo[4,3-c]pyridine

Supplier: APOLLO SCIENTIFIC
Danger

OR300399-1G OR300399-250MG
77408-644EA 360.56 USD InStock
77408-644 77408-646
3,4-Dibromo-1H-pyrazolo[4,3-c]pyridine
3,4-Dibromo-1H-pyrazolo[4,3-c]pyridine


Formula: C₆H₃Br₂N₃
Storage Temperature: Refrigerator
MDL Number: MFCD22380430
CAS Number: 1357945-30-2
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty