Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Phenylpropionic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR2976-100G OR2976-25G OR2976-5G
77408-438EA 89.66 USD InStock
77408-438 77408-440 77408-448
2-Phenylpropionic acid 97%
2-Phenylpropionic acid


Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 260
Melting Pt: -5
Density: 1.1 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002650
CAS Number: 492-37-5
EINECS: 207-752-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty