Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Mercaptoethyl)pyrazine 96%

Supplier: APOLLO SCIENTIFIC
Danger

OR451304-100G OR451304-25G
77409-778EA 430.52 USD InStock
77409-778 77409-780
(2-Mercaptoethyl)pyrazine 96%
(2-Mercaptoethyl)pyrazine


Formula: C₆H₈N₂S
Boiling Pt: 105…110 °C
Density: 1.142 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00053160
CAS Number: 35250-53-4
UN: 2810
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty