Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium triacetoxyborohydride 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR2863-100G OR2863-250G OR2863-25G
77408-142EA 34.64 USD InStock
77408-142 77408-148 77408-150
Sodium triacetoxyborohydride 95%
Sodium triacetoxyborohydride


Formula: C₆H₁₀BNaO₆
MW: 211.94 g/mol
Melting Pt: 116…120 °C (dec.)
Density: 1.43 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012211
CAS Number: 56553-60-7
UN: 1409
ADR: 4.3 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty