Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Xylenol 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR40665-5G OR40665-100G OR40665-25G
77409-574EA 21.26 USD InStock
77409-574 77409-570 77409-572
2,3-Xylenol 98%
2,3-Xylenol


Formula: C₈H₁₀O
Boiling Pt: 216…218 °C
Melting Pt: 70…76 °C
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00002227
CAS Number: 526-75-0
EINECS: 208-395-3
UN: 2928
ADR: 6.1+8,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty