Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Iodophenyl isothiocyanate

Supplier: APOLLO SCIENTIFIC
Danger

OR315240-1G OR315240-25G OR315240-5G
77409-046EA 22.26 USD InStock
77409-046 77409-048 77409-050
2-Iodophenyl isothiocyanate
2-Iodophenyl isothiocyanate


Formula: C₇H₄INS
MW: 261.09 g/mol
Boiling Pt: 296
Melting Pt: 36…39 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00041057
CAS Number: 98041-44-2
UN: 2923
ADR: 8+6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty