Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(1-Methylpiperidin-4-yl)piperazine 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR4165-1G OR4165-5G
77409-622EA 54 USD InStock
77409-622 77409-626
1-(1-Methylpiperidin-4-yl)piperazine 98%
1-(1-Methylpiperidin-4-yl)piperazine


Formula: C₁₀H₂₁N₃
Boiling Pt: 112 °C/2 mmHg
Melting Pt: 29…32 °C
Storage Temperature: Ambient
MDL Number: MFCD01075186
CAS Number: 23995-88-2
UN: 3259
ADR: 8,II
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty