Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-1-methyl-1H-1,2,3-triazole 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR301020-1G OR301020-250MG OR301020-5G
77408-724EA 72.34 USD InStock
77408-724 77408-726 77408-728
4-Bromo-1-methyl-1H-1,2,3-triazole 98%
4-Bromo-1-methyl-1H-1,2,3-triazole


Formula: C₃H₄BrN₃
Melting Pt: 98…101 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD10696344
CAS Number: 13273-53-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty