Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Formylbenzenesulfonyl chloride 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR313065-1G OR313065-5G
77409-026EA 151.8 USD InStock
77409-026 77409-028
4-Formylbenzenesulfonyl chloride 95%
4-Formylbenzenesulfonyl chloride


Formula: C₇H₅ClO₃S
Storage Temperature: Ambient
MDL Number: MFCD01861215
CAS Number: 85822-16-8
UN: 1759
ADR: 8,III
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty