Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methyl-4-pyridylmethylamine 95+%

Supplier: APOLLO SCIENTIFIC
Danger

OR318010-10G OR318010-1G OR318010-5G
77409-106EA 268.96 USD InStock
77409-106 77409-108 77409-110
N-Methyl-4-pyridylmethylamine 95+%
N-Methyl-4-pyridylmethylamine


Formula: C₇H₁₀N₂
Boiling Pt: 86 °C/3 mmHg
Storage Temperature: Ambient
MDL Number: MFCD00066937
CAS Number: 6971-44-4
EINECS: 230-198-6
UN: 2735
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty