Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR5408-100G OR5408-500G
77410-356EA 137.54 USD InStock
77410-356 77410-358
1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid
1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid


Formula: C₁₁H₁₉NO₄
MW: 229.276 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01862877
CAS Number: 98303-20-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty