Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Tributylstannyl)-1-phenoxybenzene

Supplier: APOLLO SCIENTIFIC

OR399034-250MG OR399034-1G
77409-394EA 103.92 USD InStock
77409-394 77409-392
4-(Tributylstannyl)-1-phenoxybenzene
4-(Tributylstannyl)-1-phenoxybenzene


Formula: C₂₄H₃₆OSn
Storage Temperature: Ambient
CAS Number: 99128-02-6
UN: 2788
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty