Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Tributylstannyl)-1-chlorobenzene

Supplier: APOLLO SCIENTIFIC

OR399037-1G OR399037-250MG
77409-400EA 303.6 USD InStock
77409-400 77409-402
3-(Tributylstannyl)-1-chlorobenzene
3-(Tributylstannyl)-1-chlorobenzene


Formula: C₁₈H₃₁ClSn
Storage Temperature: Ambient
CAS Number: 24344-58-9
UN: 2788
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty